Learn More
Abcam Recombinant Human Monoacylglycerol Lipase/MGL protein, 50UG
Supplier:  Abcam AB10104550UG
Encompass_Preferred
Recombinant Human Monoacylglycerol Lipase/MGL protein is a Human Full Length protein, in the 1 to 333 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Monoglyceride lipase, MGL, HU-K5, Lysophospholipase homolog, Lysophospholipase-like, Monoacylglycerol lipase, MAGL, MGLL
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1947670