Learn More
Medchemexpress LLC Recombinant human NCAM1 (CD56) protein | 97.6% | 5 UG

Supplier: Medchemexpress LLC HYP759335UG
Recombinant human NCAM-1 (CD56) is a mammalian-expressed extracellular adhesion protein supplied as a 0.22 μm filtered PBS solution with a C-terminal His tag. It is provided at high purity and concentration for use in biochemical assays, antibody validation, and studies of cell-surface interactions, with recommended long-term storage at -80 °C.
- Expressed in HEK293 mammalian cells for native glycosylation.
- C-terminal His tag for detection and purification.
- Supplied as a 0.22 μm filtered PBS solution, pH 7.4.
- High purity by reducing SDS-PAGE (>95%).
- Provided at approximately 1.98 mg/mL concentration.
- Store at -80 °C; shipped on dry ice, avoid repeated freeze-thaw.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.