Learn More
Sino Biological Recombinant Human SAE1 Protein, Biotinylated 100
Supplier:  Sino Biological 13921HNCBB100
Encompass_Preferred
A DNA sequence encoding the human SAE1 (Q9UBE0) (Met1-Lys346) was expressed and purified. The purified protein was biotinylated in vitro.
Catalog No. 50-006-99313