Learn More
Proteintech Group Inc Recombinant Human SCF 1000ug
SDP
Supplier:  Proteintech Group Inc HZ10241000UG
Encompass_Preferred
Animal-free Recombinant Human SCF is expressed in human 293 cells as a monomeric glycoprotein with an apparent molecular mass of 35 to 45 kDa. This product is produced in a human cell expression system with serum-free, chemically defined media. Production in human 293 cells offers authentic glycosylation, contributing to stability in cell growth media and other applications. SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation, and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages.
Catalog No. 50-197-3853