Learn More
ILEX LIFE SCIENCES LLC Recombinant IL3 Rat Expressed in E coli 5 ug 5 UG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A33830005
Encompass_Preferred
Ilex Life Sciences Interleukin-3 (IL-3) Rat E coli Recombinant Protein 5 g DESCRIPTION Our bioactive Interleukin-3 (IL-3) Rat E coli Recombinant Protein is a single non-glycosylated polypeptide chain that contains 144 amino acids and has a molecular mass of 16 3 kDa SOURCE Expressed in E coli FORMULATION Sterile filtered white lyophilized (freeze-dried) powder Lyophilized from a 0 2 m filtered concentrated solution in PBS pH 7 4 PURITY 97 0% (RP-HPLC and SDS-PAGE) BIOACTIVITY The ED50 as determined by the dose-dependent stimulation of the proliferation of thymidine uptake by murine MC-9 cells is 10 ng/ml corresponding to a specific activity of 1 0 x 10 5 units/mg OTHER NAMES Interleukin 3 IL3 Mast Cell Growth Factor MCGF Hematopoietic growth factor Multipotential colony-stimulating factor Multi-CSF P-cell-stimulating factor UniProtKB P04823 USAGE Intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80213