Learn More
ILEX LIFE SCIENCES LLC Recombinant KGF Human Expressed in E coli 10 ug 10 UG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A32190010
Encompass_Preferred
Ilex Life Sciences KGF (FGF-7) Human E coli Recombinant Protein 10 g DESCRIPTION Human Keratinocyte Growth Factor (KGF) also known as Fibroblast Growth Factor 7 (FGF-7) E coli Recombinant Protein is a single non-glycosylated polypeptide chain that contains 164 amino acids and has a 18995 Dalton molecular mass FORMULATION Sterile-filtered white lyophilized (freeze-dried) powder Lyophilized from a 0 2 m filtered solution in 20mM PB pH 8 0 1M NaCl PURITY 96 0% (RP-HPLC and SDS-PAGE) BIOACTIVITY Bioactivity was determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing KGF receptors yielding an ED50 10 ng/ml corresponding to a Specific Activity of 1 0 10 5 IU/mg OTHER NAMES Human KGF Keratinocyte growth factor Human FGF-7 Human FGF7 Fibroblast growth factor 7 Heparin-binding growth factor 7 HBGF-7 USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80220