Learn More
ILEX LIFE SCIENCES LLC Recombinant KGF Human Expressed in rice 10 ug 10 UG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A312120010
Encompass_Preferred
Ilex Life Sciences KGF (FGF-7) Human Plant Recombinant Protein 10 g DESCRIPTION Our bioactive KGF (FGF-7) Human Plant Recombinant Protein is a single polypeptide chain with a molecular mass of approximately 19 2 kDa SOURCE Expressed in rice grain (Oryza sativa) FORMULATION Sterile-filtered white lyophilized (freeze-dried) powder The protein was lyophilized from a 0 2 m filtered solution containing no stabilizers PURITY (SDS-PAGE) 95% BIOACTIVITY KGF has full biological activity when compared to standards The activity is determined by the dose-dependent proliferation of HUVECs and is typically 10 ng/ml corresponding to a specific activity of 100 000 U/mg OTHER NAMES Human KGF Keratinocyte growth factor Human FGF-7 Human FGF7 Fibroblast growth factor 7 Heparin-binding growth factor 7 HBGF-7 USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80365