Learn More
Abcam Recombinant mCherry protein (His tag)
Supplier:  Abcam AB199750100UG
Encompass_Preferred
Recombinant mCherry protein (His tag) is a Full Length protein, in the 1 to 236 aa range with >97% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE and HPLC. The predicted molecular weight of ab199750 protein is 28.8 kDa. - Save time and ensure accurate results - use our mCherry protein (His tag) protein as a control
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5057