Learn More
Abcam Recombinant Mouse GFAP protein (His tag), 100UG
Supplier:  Abcam AB226309100UG
Encompass_Preferred
Recombinant Mouse GFAP protein (His tag) is a Mouse Full Length protein, in the 1 to 430 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: Glial fibrillary acidic protein, GFAP, Gfap
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2105853