Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELABSCIENCE ELABSCIENCE Recombinant Protein, Mouse, NT-proBNP, Trx Tag

Supplier:  ELABSCIENCE PDEM10011120UG

Encompass_Preferred

Brain-type Natriuretic Peptide (BNP) is a nonglycosylated peptide primarily produced by ventricular myocytes and belongs to the natriuretic peptide family. The proteolytic cleavage of the 12 kDa BNP precursor results in N-terminal Pro BNP (NT-proBNP) and mature BNP. N-terminal proB-type natriuretic peptide (NT-proBNP) is a useful marker for heart failure (HF) and is thought to be secreted mainly from the ventricle. Elevated serum NT-proBNP levels are also observed in conditions such as atrial fibrillation (AF) and atrial septal defect in patients without HF.

  • Species: Mouse
  • Expression host: E.coli
  • Calculated molecular weight: 25.5 kDa
  • Observed molecular weight: 30 kDa
  • Tag: N-Trx
  • Purity: > 95%.
  • Endotoxin: < 10 EU/mg.
  • Form: Lyophilized powder
  • Storage: Stable at -20°C to -80°C for 12 months.

Catalog No. NC3987852


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.