Learn More
ILEX LIFE SCIENCES LLC Recombinant Osteoprotegerin Human Expressed in E coli 50 ug 50 UG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A31770050
Encompass_Preferred
Ilex Life Sciences Osteoprotegerin (OPG) Human E coli Recombinant Protein 50 g DESCRIPTION Our bioactive Osteoprotegerin (OPG) Human E coli Recombinant Protein is a single non-glycosylated polypeptide chain that contains 174 amino acids and has a molecular mass of 20 kDa SOURCE Expressed in E coli FORMULATION Sterile filtered white lyophilized (freeze-dried) powder The OPG was lyophilized from a 0 2 m filtered concentrated (0 5 mg/ml) solution in PBS pH 7 4 PURITY 95 0% as determined by RP-HPLC and SDS-PAGE BIOACTIVITY The activity is determined by its ability to neutralize the stimulation of U937 cells treated with 10 ng/ml of soluble RANKL corresponding to a specific activity of 100 000 IU/mg OTHER NAMES Tumor necrosis factor receptor superfamily member 11B (TNFRSF11B) Osteoclastogenesis inhibitory factor OCIF UniProtKB P05231 USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80322