Learn More
Active Motif Recombinant PKM2 protein
SDP
Supplier:  Active Motif 81302
Encompass
Full length human PKM2 protein was expressed in E. coli cells (accession number NP_872270.1) with a C-terminal 6×His-Tag. The molecular weight of the protein is 60.1 kDa. For more complete and up-to-date information, please visit the Active Motif website.
Catalog No. NC1848723