Learn More
ACROBiosystems Human NKG2D / CD314 Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems NKDH5259100UG
Encompass_Preferred
Recombinant Protein;100UG;Human NKG2D, Mouse IgG2a Fc Tag (NKD-H5259) is expressed from human 293 cells (HEK293). It contains AA Ile 73 - Val 216 (Accession # P26718-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 43.5 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human MICA, His Tag (Cat. No. MIA-H5221) at 1 μg/mL (100 μL/well) can bind Human NKG2D, Mouse IgG2a Fc Tag (Cat. No. NKD-H5259) with a linear range of 0.02-0.313 μg/mL (QC tested).
Catalog No. 50-201-9721