Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) Papain-like Protease Protein, His Tag
Supplier:  ACROBiosystems PAEC5148100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 Papain-like Protease, His Tag (PAE-C5148) is expressed from E.coli cells. It contains AA Glu 746 - Lys 1060 (Accession # YP_009725299.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 37.6 kDa. The protein migrates as 35-37 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-201-9753