Learn More
ACROBiosystems Human CD3 epsilon & CD3 delta Heterodimer Protein, His Tag&Tag Free (MALS verified)
Supplier:  ACROBiosystems CDDH52W11MG
Encompass_Preferred
Recombinant Protein;1MG;Human CD3E&CD3D Heterodimer Protein, His Tag&Tag Free (CDD-H52W1) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) & P04234-1 (CD3D)).;Human CD3E&CD3D Heterodimer Protein, His Tag&Tag Free is produced by co-expression of CD3E and CD3D, has a calculated MW of 16.6 kDa (CD3E) and 13.4 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3D contains no tag. The reducing (R) heterodimer protein migrates as 25-30 kDa (CD3E) and 20 kDa and 22 kDa (CD3D) due to glycosylation.;Immobilized Human CD3E&CD3D Heterodimer Protein, His Tag&Tag Free (Cat. No. CDD-H52W1) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3) (Cat. No. CDE-M120a) with a linear range of 0.2-6 ng/mL (QC tested).;Immobilized Human CD3E&CD3D Heterodimer Protein, His Tag&Tag Free (Cat. No. CDD-H52W1) at 1 μg/mL (100 μL/well) can bind UC
Catalog No. 50-201-9701