Learn More
ACROBiosystems PE-Labeled Human CD19 (20-291) Protein, His Tag
Supplier:  ACROBiosystems CD9HP2H3200TESTS
Encompass_Preferred
Recombinant Protein;200TESTS;PE-Labeled Human CD19 (20-291), His Tag (Cat. No. CD9-HP2H3) is produced via conjugation of PE to Human CD19 (20-291), His Tag (Cat. No. CD9-H52H2) under optimal conditions. Human CD19 (20-291), His Tag is expressed in HEK293 cells containing AA Pro 20 - Lys 291 (Accession # P15391-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.7 kDa. ;1e6 of the anti-CD19 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human CD19 (20-291), His Tag (Cat. No. CD9-HP2H3). PE Streptavidin was used as negative control (QC tested).
Catalog No. 50-201-9659