Learn More
Medchemexpress LLC Recombinant rat B7-H4 protein | >95.0% | 5 UG
SDP
Supplier:  Medchemexpress LLC HYP754855UG
Encompass_Preferred
Recombinant rat B7-H4 protein expressed in HEK293 cells with a C-terminal His tag. Designed for immunology research to evaluate B7-H4-mediated effects on T-cell activation, proliferation, and cytokine production. Supplied as a purified recombinant protein suitable for assays and binding studies.
  • Rat-derived B7-H4 protein expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Purified to greater than 95.0% as determined by SDS-PAGE.
  • Suitable for ELISA, binding studies, and functional assays.
  • Approximately 228 amino acids; 42-65 kDa apparent molecular weight.
  • Available in small research quantities (for example, 5 μg).
Catalog No. 50-002-93337