Learn More
Medchemexpress LLC Recombinant rat interleukin-21 protein | >95.0% | 2 UG

Supplier: Medchemexpress LLC HYP748182UG
Recombinant rat interleukin-21 protein supplied lyophilized for research use. Expressed in E. coli and corresponding to residues P25-S146, it is suitable for functional assays, receptor binding studies, and cell-culture experiments.
- Greater than 95.0% purity by reducing SDS-PAGE.
- Lyophilized from 0.22 μm filtered PBS, pH 7.4.
- Predicted molecular mass 14.5 kDa (apparent ~16 kDa on SDS-PAGE).
- Functional activity measured by ELISA; ED50 = 12.16 ng/mL.
- Store at -20°C; aliquot and avoid repeated freeze-thaw.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.