Learn More
Akron Biotechnology LLC Recombinant Human EPO Liquid Solution, GMP Grade, 2000 IU Vial, AK9941-0002
Supplier:  Akron Biotechnology LLC AK99410002
Encompass
Akron’s Recombinant Human Erythropoietin (rHu EPO) Solution is manufactured following all relevant cGMP guidelines for ancillary materials.Product Features:Active Substance• Amino acid sequence identical to Procrit and Epogen (epoetin alfa)• Expressed in CHO cells• All raw materials are compliant, controlled, and traceable under Akron’s Quality Management System (QMS)Manufacturing• Type II eCTD MF (#027160) on file with FDA• Animal-free media components• Formulated with a carrier protein, HSA (injectable)• Tag-free pharmaceutical biosimilar processing• Sterile filtration and aseptic fillingStability• 24-month shelf life• Store at 2-8 °C• Transport with cold packsFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications only. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.
Catalog No. NC2135670