Learn More
Abcam Recombinant S. cerevisiae KEX2 protease protein (Active), 50UG
Supplier:  Abcam AB9655450UG
Encompass_Preferred
Recombinant S. cerevisiae KEX2 protease protein (Active) is a Saccharomyces cerevisiae S288C Fragment protein, in the 110 to 667 aa range, expressed in BTI-TN-5B1-4, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC, FuncS.
Protein/ Peptide/ Enzyme Synonyms: QDS1, YNL238W, N1122, KEX2, Kexin, Protease KEX2, Proteinase YSCF
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-264-8893