Learn More
Active Motif Recombinant SIRT5 protein
SDP
Supplier:  Active Motif 31531
Encompass
Recombinant Human SIRT5 protein is useful for protein deacetylase, demalonylase, desuccinylase and deglutarylase assays and the study of enzyme kinetics, screening inhibitors and selectivity profiling.
Catalog No. NC1847507