Learn More
Active Motif Recombinant SP1 protein
SDP
Supplier:  Active Motif 81181
Encompass
Recombinant human SP1 protein was expressed in a baculovirus expression system as the full-length protein (accession number NP_612482.2) with an N-terminal FLAG tag. The molecular weight of the protein is 82.3 kDa.
Catalog No. NC1851941