Learn More
ILEX LIFE SCIENCES LLC Recombinant TGF beta 3 Human Expressed in Nicotiana tabacum chloroplast 100 ug 100 UG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC U00010100
Encompass_Preferred
TGF-beta 3 Human Plant Chloroplast-Expressed Recombinant Protein 100 g (0 1 mg) DESCRIPTION Recombinant Human TGFB3 is a non-glycosylated 112 aa (aa 301-412) disulfide-linked homodimer with a 25 425 2 Da molecular mass TGF-beta 3 Human is expressed using a novel Nicotiana tabacum chloroplast expression system eliminating contamination risk from endotoxins or animal pathogens ACCESSION NO PI0600 PURITY (SDS-PAGE) 95% ENDOTOXIN 0 0025 EU/ g protein APPEARANCE Lyophilized white powder Includes an optimal buffer for reconstitution BIOLOGICAL ACTIVITY (EC50) 0 1085 ng/mL Bioactivity was determined using the TGF -Responsive SBE Luciferase Reporter HEK293 Cell Line SHIPPING Ambient temperature STORAGE 80 C avoid repeated freeze-thaw cycles USAGE For laboratory research use only Not for use in diagnostic or therapeutic procedures SYNONYMS Transforming Growth Factor beta 3 Transforming Growth Factor- 3 TGFB3 TGF-B3 TGF- 3 TGF 3 TGF-beta-3 TGF- -3
Catalog No. 50-005-80249