Learn More
Sigma Aldrich Fine Chemicals Biosciences Recombinant Trypsin Proteo
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EMS00064X100UG
Encompass
Trypsin is a major proteolytic enzyme synthesized as a preproenzyme by pancreas and is stored as proenzyme trypsinogen in secretory granules. Trypsin belongs to the family of serine proteases that are characterized by the catalytic triad His57 Asp102 and Ser195. Trypsin is routinely used in proteomics research for peptide mapping and protein sequence work due to its highly specific cleavage resulting in a limited number of tryptic peptides. Trypsin is a pancreatic serine endoprotease which hydrolyzes peptide bonds specifically at the carboxyl side of arginine and lysine residues. The rate of hydrolysis is slower if an acidic residue is on either side of the cleavage site and cleavage may not occur if a proline residue is on the carboxyl side. The enzyme also exhibits esterase and amidase activities. Trypsin has an average molecular mass of 23.29 kDa and a pH optimum near 8.0. This product is prepared from recombinant trypsin porcine sequence. It is naturally devoid of chymotryptic a
Catalog No. 50-290-4853