Learn More
ILEX LIFE SCIENCES LLC Recombinant Uteroglobin Human Expressed in E coli 10 ug 10 UG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A37430010
Encompass_Preferred
Ilex Life Sciences Uteroglobin (CC16) Human E coli Recombinant Protein 10 g DESCRIPTION Our bioactive Uteroglobin (CC16) Human E coli Recombinant Protein is a single non-glycosylated polypeptide chain it contains 70 amino acids and has a 15 8 kDa molecular mass SOURCE E coli expressed FORMULATION Sterile filtered white lyophilized (freeze-dried) powder Lyophilized from a 0 2 m filtered concentrated solution in PBS pH 7 4 PURITY 98 0% (RP-HPLC and SDS-PAGE) BIOACTIVITY The ED50 as determined by the ability of the immobilized protein to support the adhesion of the A549 human lung carcinoma cells is 5 0 g/ml corresponding to a specific activity of 200 IU/mg OTHER NAMES Club cell protein Club cells 10 kDa secretory protein (CC10) Secretoglobin family 1A member 1 (SCGB1A1) CCPBP Urinary protein 1 (UP-1 UP1 Urine protein 1) CCSP UGB UniProtKB P11684 USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80419