Learn More
ILEX LIFE SCIENCES LLC Recombinant Uteroglobin Mouse Expressed in E coli 10 ug 10 UG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A37460010
Encompass_Preferred
Ilex Life Sciences Uteroglobin (CC16) Mouse E coli Recombinant Protein 10 g DESCRIPTION Our bioactive Uteroglobin (CC16) Mouse E coli Recombinant Protein is a non-glycosylated disulfide-linked homodimeric protein that contains 2 x 75 amino acids it has a 16 7 kDa molecular mass SOURCE E coli expressed FORMULATION Sterile filtered white lyophilized powder Lyophilized from a 0 2 m filtered concentrated solution in PBS pH 7 4 PURITY 98 0% (RP-HPLC and SDS-PAGE) BIOACTIVITY The ED50 as determined by the ability of the immobilized protein to support the adhesion of the A549 human lung carcinoma cells is 5 0 g/ml corresponding to a specific activity of 200 IU/mg OTHER NAMES Murine Club cell protein Club cells 10 kDa secretory protein (CC10) Secretoglobin family 1A member 1 (SCGB1A1) CCPBP Urinary protein 1 (UP-1 UP1 Urine protein 1) CCSP UGB UniProtKB Q06318 USAGE Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80350