Learn More
ILEX LIFE SCIENCES LLC Recombinant Uteroglobin Rat Expressed in E coli 10 ug 10 UG 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC A38200010
Encompass_Preferred
Ilex Life Sciences Uteroglobin (CC16) Rat E coli Recombinant Protein 10 g DESCRIPTION Our bioactive Uteroglobin (CC16) Rat E coli Recombinant Protein is a non-glycosylated homodimeric protein that contains 2 x 77 amino acids it has a 17 0 kDa molecular mass FORMULATION E coli expressed Sterile filtered white powder lyophilized from a 0 2 m filtered concentrated solution in PBS pH 7 4 PURITY 97 0% (RP-HPLC SDS-PAGE) BIOACTIVITY Rat CC16 recombinant is fully biologically active compared to standard The ED50 as determined by the ability of the immobilized protein to support adhesion of A549 human lung carcinoma cells is 5 0 g/ml corresponding to specific activity of 200 IU/mg OTHER NAMES Club cell protein Club cells 10 kDa secretory protein (CC10) Secretoglobin family 1A member 1 (SCGB1A1) CCPBP Urinary protein 1 (UP-1 UP1 Urine protein 1) CCSP UGB UniProtKB P17559 USAGE Lab Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80392