Learn More
Abcam Recombinant Yellow fluorescent protein
Supplier:  Abcam AB285706100UG
Encompass_Preferred
Recombinant Yellow fluorescent protein is a Full Length protein, expressed in Escherichia coli, with >97% purity and < 1 EU/µg endotoxin level.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5580