Learn More
ACROBiosystems ACROBiosystems Recombinant Fibronectin Fragment, Premium Grade (FIN-H5116) 500 μg
Supplier: ACROBiosystems FINH5116500UG
ACROBiosystems Recombinant Fibronectin fragment, premium grade (FIN-H5116) is a high-purity, E. coli-expressed human fibronectin fragment supplied as a 0.2 μm filtered solution formulated for cell culture and research applications. It contains two fibronectin segments (Pro 1361-Ser 1637 and Ala 1812-Thr 2107), is tested for sterility and endotoxin, and supports mesenchymal stem cell attachment and proliferation.
- Size: 500 μg.
- Expression system: E. coli.
- Contains amino acid segments Pro 1361-Ser 1637 and Ala 1812-Thr 2107.
- Formulation: 12.5 mM citric acid, pH 6.2, with glycerol; 0.2 μm filtered solution.
- Storage: Store at -20°C and avoid repeated freeze-thaw cycles.
- Quality control: Sterility tested and endotoxin tested.
- Applications: Promotes mesenchymal stem cell attachment and proliferation; supports stemness maintenance.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.