Learn More
Sartorius Recombumin Elite, 10% (w/v), 50 mL, Glass vial, Salt-based, Human and animal origin free, ICH Q7 cGMP
Supplier:  Sartorius 205005
Encompass
Recombumin Elite, 10% (w/v), 50 mL, Glass vial, Salt-based, Human and animal origin free, ICH Q7 cGMP
Catalog No. NC3728079