Learn More
HARVARD BIOSCIENCE INC RECORDING CHAMBER RC-26G
Supplier:  HARVARD BIOSCIENCE INC 640235
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 2.The RC-26 and RC-26G chambers are designed to be used with cells cultured onto glass coverslips, or with tissue and brain slice samples. The broad imaging area of these chambers (12 x 25 mm and 15 x 24 mm for the RC-26 and RC-26G, respectively) provides excellent access for both electrodes and water-immersion objectives. The diamond shaped geometry facilitates laminar solution flow throughout the bath area.
Catalog No. NC9646715