Learn More
Essendant Americo Buffing Pads, 13" Diameter, Red, 5/Carton
Supplier:  Essendant AM7404413
Encompass_Preferred
This pad is designed for light duty cleaning and buffing It will remove light scuff marks and dirt while producing a high gloss shine For use with standard electric floor machines orbital scrubbers and automatic scrubbers Ideal for use on machines speeds up to 800 RPM Floor Pads Application- Buffing Diameter- 13 Color(s)- Red Color Family- Red
Catalog No. 50-000-22787