Learn More
Electron Microscopy Sciences Red Tissue Marking Dye Refill 8 OZ
SDP
Supplier:  Electron Microscopy Sciences 63020RD
Encompass_Preferred
Fast drying dye is ideal for marking tissue biopsies and pathology specimens and can be used with frozen or fixed tissues. It is a specially formulated binding pigment that evenly coats the tissue without penetrating the tissue surface. The marking dyes do not interfere with diagnostic interpretation due to the fact colors are not similar to standard histopathology dyes. It can be used to permanently mark the margins of excised surgical specimens for exact orientation.
Catalog No. 50-293-42