Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc RED FIXABLE VIABILITY DYE KIT

Supplier:  Cytek Biosciences Inc R760008

Encompass_Preferred

  • Amin-reactive, fixable viability dye for flow cytometry applications.
  • Discriminates between live and dead cells.
  • Impenetrable to live cells; enters only cells with compromised membranes.
  • Maintains fluorescent signal after sample fixation.
  • Allows for subsequent intracellular staining and long-term analysis.
  • Lyophilized format for stability and convenience.
  • Compatible with various flow cytometry protocols.
  • Requires 1000 µL DMSO for reconstitution.
  • Store unreconstituted kits at -35°C to -10°C.
  • Usage for 1-2 million cells per assay.
  • Volume per assay: 5 µL of diluted dye.
  • Incubation time: 20 minutes at room temperature in darkness.
  • Storage of reconstituted dye at ≤ -20°C in a dry, dark location for up to 1 month.
  • Avoid repeated freeze-thaw cycles.
  • Compatibility with fix/perm protocols and full spectrum systems.

Catalog No. NC1997044


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.