Learn More
Medchemexpress LLC Redaporfin | 1224104-08-8 | 99.4% | 1135.11 | 5 MG

Supplier: Medchemexpress LLC HY176445MG
Redaporfin is a potent photosensitizer that causes direct antineoplastic effects and indirect immune-dependent destruction of malignant lesions. Photodynamic therapy with Redaporfin (5μM) reduces the abundance of several Golgi apparatus proteins (such as GBF1, GOLGA2, and GALT1) and ER proteins (including EIF2AK3 and PDIA3) in human osteosarcoma U2OS cells, without significantly decreasing mitochondrial import receptor subunit TOM20 homolog or β-actin.
- CAS number: 1224104-08-8
- Molecular weight: 1135.11
- Purity: 99.44%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.