Learn More
Metrohm USA REDOX BUFFER 250MV
SDP
Supplier:  Metrohm USA 62306020
Encompass
Redox standard (with Ag/AgCl/c(KCl)=3 mol/L reference electrode gives U = 250 +/-5 mV (20� C); can also be used as buffer pH=7)
Catalog No. NC0057399