Learn More
Cayman Chemical Reduced HaloperIdol 50mg
SDP
Supplier:  Cayman Chemical 2801750 MG
Encompass_Preferred
An active metabolite of haloperidol; formed via reduction of haloperidol by ketone reductase; inhibits radioligand binding to sigma-1 and dopamine D2 receptors (Kis = 1.4 and 31 nM, respectively); stimulates BDNF secretion from CCF-SSTG1 and U87MG astrocytic glial cells; inhibits norepinephrine, dopamine, and 5-HT reuptake (Kis = 21, 25, and 33 μM, respectively, in COS-7 cells expressing the human transporters); increases latency to paw withdrawal in mouse models of capsaicin-induced mechanical hypersensitivity at 0.5 mg/kg
Catalog No. 50-294-1882