Learn More
New Jersey Fluid Systems LLC REDUCER 1/16" X 1/8"
Supplier:  New Jersey Fluid Systems LLC SS100R2
Encompass
Swagelok Tube Fitting Reducer. 1/16 in. OD Swagelok tube fitting x 1/8 in. OD Tube Stub. 316 Stainless Steel
Catalog No. NC0634300