Learn More
Foxx Life Sciences Polyethylene Barbed Union Tube Fittings, Reducing Connector, 1/2 Inch x 1/4 Inch, 10/PK
Supplier:  Foxx Life Sciences 25P2501OEM
Encompass_Preferred
25P-2501-OEM: Connector, Reducer, Polyethylene, 1/2" Hose Barb x 1/4" Hose Barb, 10/pk. Foxx Life Sciences connectors are available in a variety of standard sizes. They provide a secure, watertight connection. These connectors make it possible to connect two hoses, securely and efficiently. Made from polyethylene.
Catalog No. 50-117-8749