Learn More
Grainger Reducing Adapter 316 SS 3/4 in NPT
Supplier:  Grainger 14M047
Encompass_Preferred
The Parker Reducing Adapter is made of 316 stainless steel and has female NPT threads on both ends. It reduces pipe size from 1/2" to 1/4". The maximum pressure rating is 3000 psi at 100°F. The adapter body has a hex shape for easy wrenching. It is made in the USA and is commonly used in hydraulic, pneumatic, lubrication, and industrial applications for reducing line sizes. The stainless steel material offers good corrosion resistance. This adapter allows stepping down from larger to smaller line sizes.
Catalog No. 50-275-2265