Learn More
Grainger Reducing Adapter 316 SS 3/4 x 1/4 in
Supplier:  Grainger 14M044
Encompass_Preferred
The Parker Reducing Adapter is made of 316 stainless steel and has NPT thread connections. It reduces pipe size from 1/2" to 1/4". The maximum pressure rating is 3000 psi at 100°F. The adapter body has a hex shape for easy wrench tightening. It can be used with hydraulic and pneumatic systems using petroleum-based fluids. The adapter is made in the USA.
Catalog No. 50-275-3111