Learn More
Grainger Reducing Adapter 316 SS 3/4 x 3/8 in
Supplier:  Grainger 14M045
Encompass_Preferred
The Parker Reducing Adapter is made of 316 stainless steel and has NPT thread connections. It reduces pipe size from 1/2" to 1/4". The maximum pressure rating is 3000 psi at 100°F. The adapter has a hex body for wrench tightening and is manufactured in the USA. This adapter allows transitioning to a smaller pipe size while maintaining high pressure capabilities. The stainless steel material makes it suitable for corrosive applications. With a temperature range of -20°F to 800°F, this adapter can handle a wide variety of media. The NPT connections provide a tapered thread seal when properly tightened.
Catalog No. 50-275-2578