Learn More
Grainger Reducing Bushing 316 SS 3/4 x 3/8 in
Supplier:  Grainger 14M055
Encompass_Preferred
The Parker Reducing Bushing is made from 316 stainless steel and has NPT thread connections. It reduces pipe size from 1" to 3/4". The bushing has a pressure rating of 6000 psi at 100°F. It can be used with Schedule 80 pipe and is manufactured in the USA. The bushing measures 1.81" in length and weighs 0.19 lbs.
Catalog No. 50-275-1861