Learn More
Bellco Glass, Inc. Reducing Couplers, 1/8x1/16" Single-Barbed
SDP
Supplier:  Bellco Glass, Inc. 780800013
Encompass_Preferred
Clear poly-carbonate singled barbed tube fitting. Tubing use with PVC polyurethane with a hardness of shore A 45-90. These poly-carbonate single barbed fitting are transparent so you can see that is flowing through your line. Single barbed fitting minimize leaks by having a smoother sealing surface than multi barbed fittings. Poly-carbonate has high impact strength use with air, water, and food beverage material meets FDA CFR21 1771580. Can be sterilized with steam autoclaving
Catalog No. 50-194-6386