Learn More
Grainger Reducing Tee 3/4 x1/2x1/2 In CTS PEX Wht
Supplier:  Grainger 16T748
Encompass_Preferred
The 3/4" x 1/2" x 1/2" Reducing Tee PEX Fitting by Apollo is a white PEX plastic pipe fitting used to connect one 3/4" pipe to two 1/2" pipes in a tee configuration. It has a maximum pressure rating of 100 PSI at 73°F and is NSF-pw certified for use with potable water. The fitting body and nuts are lead-free brass. It can be used for hot and cold potable water applications. This reducing tee fitting helps provide secure connections in residential and commercial plumbing and hydronic heating systems.
Catalog No. 50-276-5255