Learn More
Medchemexpress LLC Refametinib | 923032-37-5 | 99.8% | 572.34 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14691100MG
Encompass_Preferred
Refametinib (BAY 869766; RDEA119) is an orally available, potent, non-ATP-competitive, selective, allosteric MEK1/MEK2 inhibitor. It selectively binds directly to an allosteric pocket in the MEK1/2 enzymes and potently inhibits MEK activity in enzyme inhibition assays.
  • Potent MEK1/MEK2 inhibitor with IC50s of 19 nM for MEK1 and 47 nM for MEK2
  • Active in human tumor xenograft models
  • Well tolerated within therapeutic exposure range in animals
Catalog No. 50-003-63559