Learn More
TARGETMOL CHEMICALS INC REFAMETINIB R ENANTIOMER 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T126975MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Refametinib R enantiomer (RDEA119 R enantiomer) is an MEK inhibitor with an EC50 of 2.0-15 nM.Refametinib (R enantiomer) has anticancer activity and can be used in cancer research. purity: 99%
Catalog No. 50-258-1430