Learn More
Aqua Solutions Reference Fuel A, Isooctane 100%
SDP
Supplier:  Aqua Solutions SPE434
Encompass_Preferred
Melting point/Melting range: -107 °C (-160.6 °F), Boiling point/Boiling range: 98-99 °C (208.4-210.2 °F), Flash point: -12 °C (10.4 °F)
Catalog No. NC1588661