Learn More
Grainger Reflective Numbers And Letters Z PK25
Supplier:  Grainger 14R244
Encompass_Preferred
The Brady Letter Label is a 2 1/2 inch character height label with a white background and black text. It is made of vinyl material and has an adhesive backing for application. The labels are useful for identifying wires, panels, components, equipment, and more. They feature block lettering for high visibility and come in a pack of 250 labels per roll. The labels resist chemicals, abrasion, and temperatures up to 176°F.
Catalog No. 50-275-2663