Learn More
Caron Products And Services Refrigerated Incubator, 2-Door, 50 cu. ft., Temp, No RH
Supplier:  Caron Products And Services 7001501
Encompass_Preferred
50 ft3 (1,416 L) Refrigerated Incubator offers precise temperature control (5C to 70C). Ideal for a wide range of applications. This reach-in unit delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.
Catalog No. NC1855384
Add to Cart
Request a Quote